(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥96%, ee 99%

Supplier: Ambeed
Danger

Synonyms: (R,R)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane, (R,R)-2,2-Isopropylidenebis(4-phenyl-2-oxazoline), (R,R)-2,2'-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)

A468940-100MG A468940-1G A468940-250MG A468940-5G
77291-848EA 20.6 USD
77291-848 77291-850 77291-852 77594-480
(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥96%, ee 99%
(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O₂
MW: 334.42 g/mol
MDL Number: MFCD00192244
CAS Number: 150529-93-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR