Zinc(II) (R,E)-12-hydroxyoctadec-9-enoate 97%

Supplier: Ambeed
Warning

A468799-500G A468799-1000G A468799-100G
77241-210EA 11.33 USD
77241-210 77617-030 77617-032
Zinc(II) (R,E)-12-hydroxyoctadec-9-enoate 97%
Zinc(II) (R,E)-12-hydroxyoctadec-9-enoate

Precaución: for Research Use Only

Formula: C₃₆H₆₆O₆Zn
MW: 660.31 g/mol
CAS Number: 29704-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR