Fatostatin 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Propylpyridin-4-yl)-4-(p-tolyl)thiazole

A468679-100MG A468679-10MG A468679-25MG A468679-50MG A468679-5MG A468679-1MG
77265-316EA 219.39 USD
77265-316 77265-318 77265-320 77265-322 77265-324 77617-028
Fatostatin 98%
Fatostatin

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂S
MW: 294.41 g/mol
MDL Number: MFCD02376000
CAS Number: 125256-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR