Methyl trityl-D-serinate 97%

Supplier: Ambeed
Warning

A468678-1G A468678-250MG A468678-5G A468678-25G
77241-204EA 15 USD
77241-204 77241-206 77241-208 77568-924
Methyl trityl-D-serinate 97%
Methyl trityl-D-serinate

Precaución: For research use only.

Formula: C₂₃H₂₃NO₃
MW: 361.43 g/mol
MDL Number: MFCD29055252
CAS Number: 116457-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR