(2S,3R)-2-Amino-3-hydroxybutanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Thr-OH·HCl

A468662-1G A468662-25G A468662-5G A468662-100G
77265-310EA 8.24 USD
77265-310 77265-312 77265-314 77748-964
(2S,3R)-2-Amino-3-hydroxybutanoic acid hydrochloride 97%
(2S,3R)-2-Amino-3-hydroxybutanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO₃
MW: 155.58 g/mol
MDL Number: MFCD03791089
CAS Number: 82650-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR