3,5-Dimethylisoxazole-4-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A468646-1G A468646-10G
77291-846EA 11.33 USD
77291-846 77572-474
3,5-Dimethylisoxazole-4-sulfonyl chloride 98%
3,5-Dimethylisoxazole-4-sulphonyl chloride

Precaución: for Research Use Only

Formula: C₅H₆ClNO₃S
MW: 195.62 g/mol
MDL Number: MFCD00052554
CAS Number: 80466-79-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR