2'-Hydroxy-3'-nitro-[1,1'-biphenyl]-3-carboxylic acid 98+%

Supplier: Ambeed
Warning

A468553-1G A468553-25G A468553-5G
77265-304EA 8.24 USD
77265-304 77265-306 77265-308
2'-Hydroxy-3'-nitro-[1,1'-biphenyl]-3-carboxylic acid 98+%
2'-Hydroxy-3'-nitro-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉NO₅
MW: 259.21 g/mol
MDL Number: MFCD11558889
CAS Number: 376591-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR