HDM-201 99%

Supplier: Ambeed
Warning

Synonyms: Siremadlin, (S)-5-(5-Chloro-1-methyl-2-oxo-1,2-dihydropyridin-3-yl)-6-(4-chlorophenyl)-2-(2,4-dimethoxypyrimidin-5-yl)-1-isopropyl-5,6-dihydropyrrolo[3,4-d]imidazol-4(1H)-one

A468171-10MG A468171-1MG A468171-5MG A468171-100MG A468171-50MG A468171-250MG
77265-298EA 84 USD
77265-298 77265-300 77265-302 77617-026 77748-960 77748-959
HDM-201 99%
HDM-201

Precaución: for Research Use Only

Formula: C₂₆H₂₄Cl₂N6O₄
MDL Number: MFCD30502893
CAS Number: 1448867-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR