[1,1':4',1''-Terphenyl]-2',4,4'',5'-tetracarboxylic acid 98%

Supplier: Ambeed
Warning

A468092-250MG A468092-5G A468092-1G
77265-294EA 21 USD
77265-294 77265-296 77748-958
[1,1':4',1''-Terphenyl]-2',4,4'',5'-tetracarboxylic acid 98%
[1,1':4',1''-Terphenyl]-2',4,4'',5'-tetracarboxylic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₄O₈
CAS Number: 115101-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR