N1-(3-Bromobenzyl)-N1-ethylethane-1,2-diamine 97%

Supplier: Ambeed
Danger

Synonyms: NMDAR/TRPM4-IN-2 free base

A468009-250MG A468009-1G
77241-200EA 287.37 USD
77241-200 77746-957
N1-(3-Bromobenzyl)-N1-ethylethane-1,2-diamine 97%
N1-(3-Bromobenzyl)-N1-ethylethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇BrN₂
MW: 257.17 g/mol
MDL Number: MFCD21096961
CAS Number: 1353979-43-7
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR