5-Chloro-N-(4-nitrophenyl)pentanamide 97%

Supplier: Ambeed
Warning

A467696-1G A467696-5G
77241-194EA 38 USD
77241-194 77241-196 77241-198
5-Chloro-N-(4-nitrophenyl)pentanamide 97%
5-Chloro-N-(4-nitrophenyl)pentanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O₃
MDL Number: MFCD11192913
CAS Number: 1039914-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR