Phenyl 2-methoxybenzoate 95%

Supplier: Ambeed
Warning

A467089-1G A467089-5G
77265-276EA 87 USD
77265-276 77265-278
Phenyl 2-methoxybenzoate 95%
Phenyl 2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00075799
CAS Number: 10268-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR