(2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%, ee 99%

Supplier: Ambeed
Danger

Synonyms: (R)-VANOL, (2R)-3,3'-Diphenyl[2,2'-binaphthalene]-1,1'-diol

A466556-100MG A466556-1G A466556-250MG A466556-5G
77253-008EA 67.98 USD
77253-008 77253-010 77253-012 77253-014
(2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%, ee 99%
(2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol

Precaución: for Research Use Only

Formula: C₃₂H₂₂O₂
MW: 438.5100 g/mol
MDL Number: MFCD08459338
CAS Number: 147702-13-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR