4,4',4''-Tris[phenyl(m-tolyl)amino]triphenylamine 95%

Supplier: Ambeed
Warning

Synonyms: N1-Phenyl-N4,N4-bis(4-(phenyl(m-tolyl)amino)phenyl)-N1-(m-tolyl)benzene-1,4-diamine, m-MTDATA

A466068-1G A466068-250MG A466068-5G
77291-830EA 78.28 USD
77291-830 77291-832 77291-834
4,4',4''-Tris[phenyl(m-tolyl)amino]triphenylamine 95%
4,4',4''-Tris[phenyl(m-tolyl)amino]triphenylamine

Precaución: for Research Use Only

Formula: C₅₇H₄₈N₄
MW: 789.04 g/mol
MDL Number: MFCD00799401
CAS Number: 124729-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR