H-Ser(Bzl)-OH.HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser(Bzl)-OH·HCl

A465881-1G A465881-100G A465881-25G A465881-5G A465881-10G
77241-192EA 9.27 USD
77241-192 77617-014 77617-016 77746-952 77746-951
H-Ser(Bzl)-OH.HCl 97%
H-Ser(Bzl)-OH.HCl

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₃
MW: 231.67 g/mol
MDL Number: MFCD20264821
CAS Number: 76614-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR