4-Isopropyl-2-nitroaniline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Nitro-4-(prop-2-yl)aniline, 4-Amino-3-nitrocumene
A465851-1G
A465851-250MG
77291-826EA
99
USD
77291-826
77291-828
4-Isopropyl-2-nitroaniline 97%
4-Isopropyl-2-nitroaniline
Precaución: for Research Use Only
Formula:
C₉H₁₂N₂O₂ |
MDL Number:
MFCD00092920 CAS Number: 63649-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...