2,5-Diiodobenzoic acid 97%

Supplier: Ambeed
Warning

A465762-100G A465762-10G A465762-1G A465762-25G A465762-5G
77252-998EA 184 USD
77252-998 77253-000 77253-002 77253-004 77253-006
2,5-Diiodobenzoic acid 97%
2,5-Diiodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄I₂O₂
MW: 373.92
MDL Number: MFCD00045801
CAS Number: 14192-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR