(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A465012-100MG A465012-1G A465012-250MG
77252-982EA 103 USD
77252-982 77252-984 77252-986
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate 98%
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂₇H₄₀F₃O₃P₂RhS
MDL Number: MFCD00269863
CAS Number: 136705-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR