6-Amino-2-bromonicotinic acid 97%

Supplier: Ambeed
Warning

A464927-100MG A464927-250MG
77265-268EA 79.31 USD
77265-268 77265-270
6-Amino-2-bromonicotinic acid 97%
6-Amino-2-bromonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD26516111
CAS Number: 1805444-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR