Salicylaldehyde ethyl ether 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxybenzaldehyde

A464795-100G A464795-10G A464795-25G A464795-5G
77265-260EA 46.35 USD
77265-260 77265-262 77265-264 77265-266
Salicylaldehyde ethyl ether 97%
Salicylaldehyde ethyl ether

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00003316
CAS Number: 613-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR