4-Iodo-3-methyl-1H-pyrazole 95%
Supplier: Ambeed
|
Warning
|
A464632-100G
A464632-10G
A464632-25G
77279-302EA
153
USD
77279-302
77279-304
77279-306
4-Iodo-3-methyl-1H-pyrazole 95%
4-Iodo-3-methyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₄H₅IN₂ |
MDL Number:
MFCD09908176 CAS Number: 15802-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...