N-(Chloromethyl)phthalimide 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Chloromethyl)isoindoline-1,3-dione

A464485-10G A464485-25G
77265-256EA 17 USD
77265-256 77265-258
N-(Chloromethyl)phthalimide 95%
N-(Chloromethyl)phthalimide

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MDL Number: MFCD00005898
CAS Number: 17564-64-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR