4-Hydroxy-7-(trifluoromethyl)quinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A464341-1G A464341-5G
77291-818EA 29.87 USD
77291-818 77291-820
4-Hydroxy-7-(trifluoromethyl)quinoline-3-carboxylic acid 95%
4-Hydroxy-7-(trifluoromethyl)quinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₆F₃NO₃
MW: 257.17 g/mol
MDL Number: MFCD00006771
CAS Number: 574-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR