4-Bromo-1,3-benzodioxole 95%

Supplier: Ambeed
Warning

A464205-100MG A464205-1G A464205-250MG A464205-5G
77291-810EA 22 USD
77291-810 77291-812 77291-814 77291-816
4-Bromo-1,3-benzodioxole 95%
4-Bromo-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₅BrO₂
MDL Number: MFCD02681890
CAS Number: 6698-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR