UNC0642 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4-Difluoropiperidin-1-yl)-N-(1-isopropylpiperidin-4-yl)-6-methoxy-7-(3-(pyrrolidin-1-yl)propoxy)quinazolin-4-amine

A464128-10MG A464128-1MG A464128-50MG A464128-5MG
77241-184EA 151 USD
77241-184 77241-186 77241-188 77241-190
UNC0642 98%
UNC0642

Precaución: for Research Use Only

Formula: C₂₉H₄₄F₂N₆O₂
MW: 546.6955 g/mol
MDL Number: MFCD28044322
CAS Number: 1481677-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR