(R)-2-Chloro-6-(2-hydroxy-3-((2-methyl-1-(naphthalen-2-yl)propan-2-yl)amino)propoxy)benzonitrile hydrochloride 98%

Supplier: Ambeed
Danger

A464000-10MG A464000-1MG A464000-50MG A464000-5MG
77241-176EA 116 USD
77241-176 77241-178 77241-180 77241-182
(R)-2-Chloro-6-(2-hydroxy-3-((2-methyl-1-(naphthalen-2-yl)propan-2-yl)amino)propoxy)benzonitrile hydrochloride 98%
(R)-2-Chloro-6-(2-hydroxy-3-((2-methyl-1-(naphthalen-2-yl)propan-2-yl)amino)propoxy)benzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₆Cl₂N₂O₂
CAS Number: 324523-20-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR