6-Borono-2-naphthoic acid 97%

Supplier: Ambeed
Warning

A463775-100MG A463775-1G A463775-250MG
77279-296EA 90.64 USD
77279-296 77279-298 77279-300
6-Borono-2-naphthoic acid 97%
6-Borono-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BO₄
MW: 216 g/mol
MDL Number: MFCD09751357
CAS Number: 2377607-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR