2,2':6',2''-Terpyridine 97%

Supplier: Ambeed
Warning

A463490-1G A463490-250MG
77252-956EA 5.15 USD
77252-956 77252-958 77252-960 77252-962 77252-964 77252-966
2,2':6',2''-Terpyridine 97%
2,2':6',2''-Terpyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₁N₃
MDL Number: MFCD00006213
CAS Number: 1148-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR