4-(Ethylamino)piperidine-4-carboxamide 95%

Supplier: Ambeed
Warning

A463401-100MG A463401-250MG
77291-802EA 41.2 USD
77291-802 77291-804
4-(Ethylamino)piperidine-4-carboxamide 95%
4-(Ethylamino)piperidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₈H₁₇N₃O
MW: 171.24 g/mol
MDL Number: MFCD00087130
CAS Number: 84100-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR