4-Octylbenzene-1,2-diol 97%

Supplier: Ambeed
Warning

A463051-100MG A463051-1G A463051-250MG
77241-168EA 110.21 USD
77241-168 77241-170 77241-172
4-Octylbenzene-1,2-diol 97%
4-Octylbenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
MDL Number: MFCD30342423
CAS Number: 7580-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR