2,4-Difluoroanisole 98%

Supplier: Ambeed
Danger

Synonyms: 2,4-Difluoro-1-methoxybenzene

A462951-25G A462951-5G
77252-950EA 43.26 USD
77252-950 77252-952
2,4-Difluoroanisole 98%
2,4-Difluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MW: 144.12 g/mol
MDL Number: MFCD00010731
CAS Number: 452-10-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR