2-Chloro-5-iodobenzonitrile 98%

Supplier: Ambeed
Danger

A462626-25G A462626-5G A462626-1G
77265-246EA 66.95 USD
77265-246 77748-942 77748-941
2-Chloro-5-iodobenzonitrile 98%
2-Chloro-5-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClIN
MW: 263.4629 g/mol
MDL Number: MFCD00672973
CAS Number: 289039-29-8
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR