Dibenzo[b,f]azocin-6(5H)-one 98%
Supplier: Ambeed
|
Warning
|
A462304-100MG
A462304-1G
A462304-250MG
77265-240EA
47.38
USD
77265-240
77265-242
77265-244
Dibenzo[b,f]azocin-6(5H)-one 98%
Dibenzo[b,f]azocin-6(5H)-one
Precaución: for Research Use Only
Formula:
C₁₅H₁₁NO MW: 221.25 g/mol |
MDL Number:
MFCD01925280 CAS Number: 23112-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...