1-Methylnaphthalen-2-ol 98%

Supplier: Ambeed
Warning

A461975-1G A461975-5G
77241-152EA 115.36 USD
77241-152 77241-154
1-Methylnaphthalen-2-ol 98%
1-Methylnaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.19 g/mol
MDL Number: MFCD00029266
CAS Number: 1076-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR