3-(Cyclopentylcarbamoyl)phenylboronic acid neopentyl glycol ester 95%

Supplier: Ambeed
Warning

Synonyms: N-Cyclopentyl-3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzamide

A461950-1G A461950-250MG A461950-5G
77279-284EA 69 USD
77279-284 77279-286 77279-288
3-(Cyclopentylcarbamoyl)phenylboronic acid neopentyl glycol ester 95%
3-(Cyclopentylcarbamoyl)phenylboronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₄BNO₃
MDL Number: MFCD06659914
CAS Number: 850567-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR