4-Acetoxybiphenyl 99%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Biphenyl]-4-yl acetate

A461686-100G A461686-25G A461686-500G A461686-5G
77291-784EA 108 USD
77291-784 77291-786 77291-788 77291-790
4-Acetoxybiphenyl 99%
4-Acetoxybiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 33
MDL Number: MFCD00014979
CAS Number: 148-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR