tert-Butyl ((3S,4S)-3-aminotetrahydro-2H-pyran-4-yl)carbamate 98%

Supplier: Ambeed
Warning

A461252-100MG A461252-250MG
77265-224EA 132.87 USD
77265-224 77265-226
tert-Butyl ((3S,4S)-3-aminotetrahydro-2H-pyran-4-yl)carbamate 98%
tert-Butyl ((3S,4S)-3-aminotetrahydro-2H-pyran-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.27 g/mol
MDL Number: MFCD30471688
CAS Number: 1802334-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR