4-Amino-2-chloropyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chlor-4-pyrimidinamin, 2-Chloro-4-pyrimidinamine, 2-Chloropyrimidin-4-amine, 2-Chloropyrimidin-4-ylamine

A461094-100G A461094-10G A461094-1G A461094-25G A461094-5G A461094-500G
77291-774EA 89 USD
77291-774 77291-776 77291-778 77291-780 77291-782 77748-934
4-Amino-2-chloropyrimidine 98%
4-Amino-2-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
MW: 129.55
MDL Number: MFCD00194051
CAS Number: 7461-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR