1,3-Di-Boc-2-(2-Hydroxyethyl)Guanidine 97%

Supplier: Ambeed
Warning

A460910-100MG A460910-1G A460910-250MG
77241-146EA 47.38 USD
77241-146 77241-148 77241-150
1,3-Di-Boc-2-(2-Hydroxyethyl)Guanidine 97%
1,3-Di-Boc-2-(2-Hydroxyethyl)Guanidine

Precaución: for Research Use Only

Formula: C₁₃H₂₅N₃O₅
MW: 303.35 g/mol
MDL Number: MFCD03791114
CAS Number: 215050-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR