3-Methylbiphenyl 95%

Supplier: Ambeed
Warning

Synonyms: 3-Phenyltoluene

A460701-1G A460701-25G A460701-5G
77279-258EA 15.45 USD
77279-258 77279-260 77616-992
3-Methylbiphenyl 95%
3-Methylbiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₂
MW: 168.24 g/mol
MDL Number: MFCD00008533
CAS Number: 643-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR