(2,3-Dihydro-1H-indol-5-ylmethyl)amine dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Indolin-5-ylmethanamine dihydrochloride
A460248-100MG
A460248-1G
A460248-250MG
A460248-5G
77241-132EA
128.75
USD
77241-132
77241-134
77241-136
77241-138
(2,3-Dihydro-1H-indol-5-ylmethyl)amine dihydrochloride 97%
(2,3-Dihydro-1H-indol-5-ylmethyl)amine dihydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₄Cl₂N₂ MW: 221.13 g/mol |
MDL Number:
MFCD16039339 CAS Number: 1242338-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...