3-[(N-Methoxy-N-methylamino)carbonyl]-5-nitrobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-[Methoxy(methyl)carbamoyl]-5-nitrophenylboronic acid

A460145-1G A460145-250MG A460145-5G
77252-924EA 100.94 USD
77252-924 77252-926 77252-928
3-[(N-Methoxy-N-methylamino)carbonyl]-5-nitrobenzeneboronic acid 98%
3-[(N-Methoxy-N-methylamino)carbonyl]-5-nitrobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BN₂O₆
MW: 254.004 g/mol
MDL Number: MFCD07783876
CAS Number: 871332-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR