3-Isobutyl-1-methylxanthine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione

A459882-1MG A459882-100MG A459882-1G
77279-252EA 5.15 USD
77279-252 77748-931 77748-932
3-Isobutyl-1-methylxanthine 98%
3-Isobutyl-1-methylxanthine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₄O₂
MW: 222.25 g/mol
MDL Number: MFCD00005584
CAS Number: 28822-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR