4-Fluoro-2-formylbenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-formylphenylboronic acid pinacol ester, 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, Benzaldehyde, 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

A459770-100MG A459770-1G A459770-250MG A459770-5G
77252-916EA 12.36 USD
77252-916 77252-918 77252-920 77252-922
4-Fluoro-2-formylbenzeneboronic acid pinacol ester 98%
4-Fluoro-2-formylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
MDL Number: MFCD18731000
CAS Number: 943310-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR