3',5'-Dimethyl-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetyl-4,6-dimethylphenol, 2'-Hydroxy-3',5'-dimethylacetophenone

A459728-100G A459728-25G A459728-5G
77279-246EA 408.91 USD
77279-246 77279-248 77279-250
3',5'-Dimethyl-2'-hydroxyacetophenone 98%
3',5'-Dimethyl-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2000 g/mol
MDL Number: MFCD01631133
CAS Number: 1198-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR