(S)-3,3'-Bis(2-naphthyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A459617-100MG
A459617-1G
A459617-250MG
77252-910EA
22
USD
77252-910
77252-912
77252-914
(S)-3,3'-Bis(2-naphthyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol ≥98%, ee 99%
(S)-3,3'-Bis(2-naphthyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol
Precaución: for Research Use Only
Formula:
C₄₀H₃₄O₂ |
CAS Number:
1121764-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...