2-Bromobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A459092-10G A459092-1G A459092-25G A459092-5G
77291-762EA 11 USD
77291-762 77291-764 77291-766 77291-768
2-Bromobenzenesulfonyl chloride 98%
2-Bromobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₄BrClO₂S
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR