2-(Piperazin-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

A458342-100MG A458342-250MG A458342-25G A458342-5G
77252-902EA 8 USD
77252-902 77252-904 77252-906 77252-908
2-(Piperazin-1-yl)acetic acid 95%
2-(Piperazin-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₂
MDL Number: MFCD03119773
CAS Number: 37478-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR