1-Methylxanthine 97%

Supplier: Ambeed
Warning

Synonyms: 3,7-Dihydro-1-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-1-methylpurine, 1-Methyl-1H-purine-2,6(3H,7H)-dione

A458316-100MG A458316-1G A458316-250MG A458316-5G
77241-112EA 44.29 USD
77241-112 77241-114 77241-116 77241-118
1-Methylxanthine 97%
1-Methylxanthine

Precaución: for Research Use Only

Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
MDL Number: MFCD00005561
CAS Number: 6136-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR