4-(5,7-Dichloro-1H-benzo[d]imidazol-2-yl)aniline 97%

Supplier: Ambeed
Warning

A458047-100MG A458047-250MG
77241-102EA 41 USD
77241-102 77241-104
4-(5,7-Dichloro-1H-benzo[d]imidazol-2-yl)aniline 97%
4-(5,7-Dichloro-1H-benzo[d]imidazol-2-yl)aniline

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₂N₃
MW: 278.13 g/mol
MDL Number: MFCD19374205
CAS Number: 303099-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR